Learn More
Sino Biological Recombinant Human LAP3 Protein (His Tag)
Supplier:  Sino Biological 15480H08H100
Encompass_Preferred
A DNA sequence encoding the human LAP3 (NP_056991.2) (Met1-Ala519) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9564