Learn More
Sino Biological Recombinant Human PKLR Protein (His Tag)
Supplier:  Sino Biological 15501H07E50
Encompass_Preferred
A DNA sequence encoding the human PKLR (NP_000289.1) (Leu47-Ser574) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9572