Learn More
Sino Biological Recombinant Human LRRC32 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 15508H08H50
Encompass_Preferred
A DNA sequence encoding the human LRRC32 (NP_005503.1) (Met1-Asn627) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9574