Learn More
Sino Biological Recombinant Human MAGOH Protein (His Tag)
Supplier:  Sino Biological 15516H08E100
Encompass_Preferred
A DNA sequence encoding the human MAGOH (NP_002361.1) (Met1-Ile146) was expressed with a polyhistide tag at the C-terminus.
Catalog No. 50-161-9575