Learn More
Cole-Parmer Fluke Calibration 1551A-9 Handheld Thermometer, Fixed RTD, –50 to 160C, 3/16x9
Catalog No. NC0284487 Shop All Cole-Parmer Products
Encompass
Whether youre verifying the accuracy of a temperature sensor or performing a procedure for the pharmaceutical industry, the Fluke Stik thermometer from Hart Scientific gives you a safe, portable, and accurate solution for all of your temperature calibration applications. The probe and digital readout are fixed together, calibrated as a system, and accurate to 0.1F (0.05C) over the full temperature range with selectable resolution of 0.1, 0.01 or 0.001. The instrument comes with an easy-to-read temperature trend indicator that you configure to meet your applications stability requirements. The indicator tells you if your temperature is trending up or down and lets you know when your temperature source is stable enough for an accurate measurement.
Catalog No. NC0284487 Supplier Cole-Parmer Supplier No. 9320207
Whether youre verifying the accuracy of a temperature sensor or performing a procedure for the pharmaceutical industry, the Fluke Stik thermometer from Hart Scientific gives you a safe, portable, and accurate solution for all of your temperature calibration applications. The probe and digital readout are fixed together, calibrated as a system, and accurate to 0.1F (0.05C) over the full temperature range with selectable resolution of 0.1, 0.01 or 0.001. The instrument comes with an easy-to-read temperature trend indicator that you configure to meet your applications stability requirements. The indicator tells you if your temperature is trending up or down and lets you know when your temperature source is stable enough for an accurate measurement.