Learn More
Sino Biological Recombinant Human Somatostatin Receptor 1/SSR1 Protein (hFc Tag)
Supplier:  Sino Biological 15526H02H100
Encompass_Preferred
A DNA sequence encoding the human SSR1 (NP_003135.2) (Met1-Thr207) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9579