Learn More
Sino Biological Recombinant Human Livin/BIRC7 Protein (His Tag)
Supplier:  Sino Biological 15527H08E100
Encompass_Preferred
A DNA sequence encoding the human BIRC7 (AAH14475.1) (Met1-Ser298) was expressed with a polyhistide tag at the C-terminus.
Catalog No. 50-161-9581