Learn More
Sino Biological Recombinant Human RBKS Protein (His Tag)
Supplier:  Sino Biological 15530H07E50
Encompass_Preferred
A DNA sequence encoding the human RBKS (NP_071411.1) (Met1-Phe322) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9583