Learn More
Sino Biological Recombinant Human PIG3 Protein (His Tag)
Supplier:  Sino Biological 15531H07E100
Encompass_Preferred
A DNA sequence encoding the human TP53I3 (NP_004872.2) (Met1-Gln332) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9584