Learn More
Sino Biological Recombinant Human MAB21L2 Protein (His & ZZ Tag)
Supplier:  Sino Biological 15540H34E100
Encompass_Preferred
A DNA sequence encoding the human MAB21L2 (NP_006430.1) (Met1-Leu359) was expressed with a N-terminal polyhistidine-tagged ZZ tag at the N-terminus.
Catalog No. 50-161-9586