Learn More
Sino Biological Recombinant Human VMO1 Protein
Supplier:  Sino Biological 15571HCCH100
Encompass_Preferred
A DNA sequence encoding the human VMO1 (NP_872372.1) (Met1-Ser202) was expressed with five amino acids (DDDDK) at the C-terminus.
Catalog No. 50-161-9597