Learn More
Sino Biological Recombinant Human Amelotin Protein (mFc Tag)
Supplier:  Sino Biological 15576H05H100
Encompass_Preferred
A DNA sequence encoding the human AMTN (NP_997722.1) (Met1-Gln209) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9601