Learn More
Sino Biological Recombinant Human INSL6 Protein (His Tag)
Supplier:  Sino Biological 15579H08H100
Encompass_Preferred
A DNA sequence encoding the human INSL6 (NP_009110.2) (Met1-Phe198) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9603