Learn More
Sino Biological Recombinant Human GSTT2B Protein (His Tag)
Supplier:  Sino Biological 15581H08H100
Encompass_Preferred
A DNA sequence encoding the human GSTT2B (NP_000845.1) (Met1-Pro244) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9605