Learn More
Sino Biological Recombinant Human CPVL Protein (His Tag)
Supplier:  Sino Biological 15590H08H100
Encompass_Preferred
A DNA sequence encoding the human CPVL (AAH16838.1) (Met1-Gly476) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9607