Learn More
Sino Biological Recombinant Human ATOX1 Protein (His Tag)
Supplier:  Sino Biological 15594H07E100
Encompass_Preferred
A DNA sequence encoding the human ATOX1 (NP_004036.1) (Met1-Glu68) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9609