Learn More
Sino Biological Recombinant Human MUSTN1 Protein (His Tag)
Supplier:  Sino Biological 15597H07E100
Encompass_Preferred
A DNA sequence encoding the human MUSTN1 (NP_995325.3) (Met1-Gly82) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9611