Learn More
Fluidigm Corp 155GD X8 ANTIBODY LABELING
SDP
Catalog No. NC0873399
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 155Gd metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC0873399 Supplier Fluidigm Corp Supplier No. 201155A
This Maxpar kit lets you conjugate IgG antibodies with the 155Gd metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.