Learn More
Sino Biological Recombinant Human FAM150A Protein (mFc Tag), HPLC-verified
Supplier:  Sino Biological 15606H05H100
Encompass_Preferred
A DNA sequence encoding the human FAM150A (NP_997296.1) (Met1-Thr129) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9619