Learn More
Sino Biological Recombinant Human OBP2B Protein (mFc Tag)
Supplier:  Sino Biological 15608H05H100
Encompass_Preferred
A DNA sequence encoding the human OBP2B (NP_055396.1) (Met1-His170) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9621