Learn More
Sino Biological Recombinant Human GABARAP Protein (His & MBP Tag)
Supplier:  Sino Biological 15622H10E100
Encompass_Preferred
A DNA sequence encoding the human GABARAP (NP_009209.1) (Met1-Leu117) was expressed with an N-terminal polyhistidine-tagged MBP tag at the N-terminus.
Catalog No. 50-161-9625