Learn More
Sino Biological Recombinant Human alpha Lactalbumin/LALBA Protein (mFc Tag)
Supplier:  Sino Biological 15631H05H100
Encompass_Preferred
A DNA sequence encoding the human LALBA (NP_002280.1) (Met1-Leu142) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9629