Learn More
Bio Rad Laboratories 1563402 NUVIA CPRIME MEDIA 100
Catalog No. NC2101489
Encompass
Nuvia cPrime Media100 ml, hydrophobic cation exchangechromatography media, 70 µm particle size
This item is not returnable. View return policy
Catalog No. NC2101489 Supplier Bio Rad Laboratories Supplier No. 1563402
Nuvia cPrime Media100 ml, hydrophobic cation exchangechromatography media, 70 µm particle size