Learn More
Sino Biological Recombinant Human RGS1 Protein (His Tag)
Supplier:  Sino Biological 15665H07E100
Encompass_Preferred
A DNA sequence encoding the human RGS1 (NP_002913.3) (Met1-Lys196) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9651