Learn More
Sino Biological Recombinant Human p67phox Protein
Supplier:  Sino Biological 15669HNCB100
Encompass_Preferred
A DNA sequence encoding the human NCF2 (AAH01606.1) (Met1-Val526) was expressed and purified with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-9655