Learn More
Sino Biological Recombinant Human BBOX1 Protein (His & GST Tag)
Supplier:  Sino Biological 15675H20B100
Encompass_Preferred
A DNA sequence encoding the human BBOX1 (O75936) (Met1-Asn387) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9664