Learn More
Sino Biological Recombinant Human P38 Gamma/MAPK12 Protein
Supplier:  Sino Biological 15676HNCB50
Encompass_Preferred
A DNA sequence encoding the human MAPK12 (P53778) (Met1-Leu367) was fused with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-9669