Learn More
Sino Biological Recombinant Human FAM3C Protein (hFc Tag)
Supplier:  Sino Biological 15678H02H100
Encompass_Preferred
A DNA sequence encoding the human FAM3C (Q92520)(Met1-Asp227) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9670