Learn More
Sino Biological Recombinant Human Sphingosine Kinase 1 Protein (His & GST Tag)
Supplier:  Sino Biological 15679H20B50
Encompass_Preferred
A DNA sequence encoding the human SPHK1 (Q9NYA1-1)(Met1-Leu384) was expressed the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9676