Learn More
Sino Biological Recombinant Human MZB1/PERP1 Protein (hFc Tag)
Supplier:  Sino Biological 15682H02H100
Encompass_Preferred
A DNA sequence encoding the human MZB1 (NP_057543.2) (Met1-Thr185) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9683