Learn More
Sino Biological Recombinant Human MZB1/PERP1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 15682H08H100
Encompass_Preferred
A DNA sequence encoding the human MGC29506 (Q8WU39-1) (Met 1-Thr 185) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9685