Learn More
Sino Biological Recombinant Human CADM2 Protein (ECD, His Tag)
Supplier:  Sino Biological 15690H08H100
Encompass_Preferred
A DNA sequence encoding the human CADM2 (NP_001161147.1) (Gly30-Asp335) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9693