Learn More
Sino Biological Recombinant Human CDNF Protein (hFc Tag)
Supplier:  Sino Biological 15691H02H100
Encompass_Preferred
A DNA sequence encoding the human CDNF (NP_001025125.2) (Met1-Leu187) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9695