Learn More
Sino Biological Recombinant Human CDNF Protein (His Tag)
Supplier:  Sino Biological 15691H08H100
Encompass_Preferred
A DNA sequence encoding the human CDNF (NP_001025125.2) (Met1-Leu187) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9697