Learn More
Sino Biological VEGF121b Protein, Human, Recombinant
Catalog No. 501619700
Encompass_Preferred
A DNA sequence encoding the human VEGF 121 isoform b (P15692-9) (Met1-Asp147,with last 6 amino acids switch from CDKPRR to SLTRKD ) was expressed.
Catalog No. 50-161-9700 Supplier Sino Biological Supplier No. 15692HNAH50
A DNA sequence encoding the human VEGF 121 isoform b (P15692-9) (Met1-Asp147,with last 6 amino acids switch from CDKPRR to SLTRKD ) was expressed.