Learn More
Sino Biological Recombinant Human Neurofascin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 15694H08H100
Encompass_Preferred
A DNA sequence encoding the human NFASC (O94856-12) (Met1-Gln939) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-9707