Learn More
Sino Biological Recombinant Human LRRTM2 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 15698H02H100
Encompass_Preferred
A DNA sequence encoding the human LRRTM2 (NP_056379.1) (Met1-Arg422) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9709