Learn More
Sino Biological Recombinant Human LRRTM2 Protein (His Tag)
Supplier:  Sino Biological 15698H08H100
Encompass_Preferred
A DNA sequence encoding the human LRRTM2 (NP_056379.1) (Met1-Arg422) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9711