Learn More
Abcam 156Gd Metal Conjugation Kit - Lightning-Link, 100UG
Catalog No. NC2270774
Encompass_Preferred
Labeling in < 4 hrs with 30 secs hands-on time using 156Gd Metal Conjugation Kit - Lightning-Link ab269838. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2270774 Supplier Abcam Supplier No. AB269838100UG
Labeling in < 4 hrs with 30 secs hands-on time using 156Gd Metal Conjugation Kit - Lightning-Link ab269838. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement