Learn More
Sino Biological Recombinant Human MUCL1 Protein (His Tag)
Supplier:  Sino Biological 15716H08H100
Encompass_Preferred
A DNA sequence encoding the human MUCL1 (NP_477521.1) (Met1-Pro90) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9713