Learn More
Sino Biological Recombinant Human PGLYRP4 Protein (His Tag)
Supplier:  Sino Biological 15730H08H200
Encompass_Preferred
A DNA sequence encoding the human PGLYRP4 (NP_065126.2) (Met1-His373) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9715