Learn More
Sino Biological Recombinant Human MCEMP1 Protein (mFc Tag)
Supplier:  Sino Biological 15757H04H100
Encompass_Preferred
A DNA sequence encoding the human MCEMP1 (NP_777578.2) (Lys107-Gln187) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-9721