Learn More
Sino Biological Recombinant Human ULBP6/RAET1L Protein (His Tag)
Supplier:  Sino Biological 15759H08H100
Encompass_Preferred
A DNA sequence encoding the human RAET1L (AAK91503.1) (Met1-Ser217) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9725