Learn More
Sino Biological Recombinant Human ANTXR2 Protein (hFc Tag)
Supplier:  Sino Biological 15762H02H100
Encompass_Preferred
A DNA sequence encoding the human ANTXR2 (NP_477520.2) (Met1-Asn317) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9727