Learn More
Sino Biological Recombinant Human EphA1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 15789H02H100
Encompass_Preferred
A DNA sequence encoding the human EPHA1 (EAL23789.1) (Met1-Glu547) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9733