Learn More
Sino Biological Recombinant Human NEU2 Protein (His Tag)
Supplier:  Sino Biological 15845H08H100
Encompass_Preferred
A DNA sequence encoding the human NEU2 (CAB41449.1) (Met1-Gln380) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9739