Learn More
Sino Biological Recombinant Human CSTL1 Protein (hFc Tag)
Supplier:  Sino Biological 15857H02H100
Encompass_Preferred
A DNA sequence encoding the human CSTL1 (AAI17397.1) (Met1-Arg145) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9741