Learn More
Sino Biological Recombinant Human IL-31RA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 15878H08H50
Encompass_Preferred
A DNA sequence encoding the human IL31RA (EAW54932.1) (Met1-Ser516) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9751