Learn More
Sino Biological Recombinant Human NPTN Protein (ECD, His Tag)
Supplier:  Sino Biological 15881H08H100
Encompass_Preferred
A DNA sequence encoding the human NPTN (NP_059429.1) (Met1-Leu221) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9752