Learn More
Sino Biological Recombinant Human ATP1B2 Protein (His Tag)
Supplier:  Sino Biological 15882H07H100
Encompass_Preferred
A DNA sequence encoding the human ATP1B2 (NP_001669.3) (Asp68-Thr290) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9754